Learn More
Trichloroacetyl chloride, 99%
Description
Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.
Solubility
Reacts violently with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, alcohols, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | -146°C |
| Density | 1.629 |
| Boiling Point | 114°C to 116°C |
| Refractive Index | 1.47 |
| Quantity | 1000 g |
| UN Number | UN2442 |
| Beilstein | 774120 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts violently with water. |
| Formula Weight | 181.83 |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H330-H335 |
| Hazard Statement | GHS H Statement H330-H314-H318-H302 Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRICHLOROACETYL CHLORIDE |
| EINECSNumber | 200-926-7 |
| RTECSNumber | AO7140000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only