missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetonitrile 98.0+%, TCI America™

Catalog No. T037225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T037225G 25 g
T0372100G 100 g
T0372500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T037225G Supplier TCI America Supplier No. T037225G

Chemical Identifiers

CAS 545-06-2
Molecular Formula C2Cl3N
Molecular Weight (g/mol) 144.379
MDL Number MFCD00001842
InChI Key DRUIESSIVFYOMK-UHFFFAOYSA-N
Synonym trichloroacetonitrile, acetonitrile, trichloro, tritox, cyanotrichloromethane, trichloromethyl cyanide, trichloroethanenitrile, trichlouracetonitril, trichlor-acetonitrile, trichloromethylnitrile, nitrile trichloracetique
PubChem CID 11011
ChEBI CHEBI:82541
IUPAC Name 2,2,2-trichloroacetonitrile
SMILES C(#N)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
UN Number 3276
Formula Weight 144.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichloroacetonitrile
EINECSNumber (2)-3608
RTECSNumber AM2450000
TSCA Yes