missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetonitrile, 98%

Catalog No. p-4509966
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139370050 5 g Glass bottle
AC139371000 100 g Glass bottle
AC139375000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC139370050 Supplier Thermo Scientific Chemicals Supplier No. 139370050
Only null left

CAS: 545-06-2 | C2Cl3N | 144.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 545-06-2
Molecular Formula C2Cl3N
Molecular Weight (g/mol) 144.39
InChI Key DRUIESSIVFYOMK-UHFFFAOYSA-N
Synonym trichloroacetonitrile, acetonitrile, trichloro, tritox, cyanotrichloromethane, trichloromethyl cyanide, trichloroethanenitrile, trichlouracetonitril, trichlor-acetonitrile, trichloromethylnitrile, nitrile trichloracetique
PubChem CID 11011
ChEBI CHEBI:82541
IUPAC Name 2,2,2-trichloroacetonitrile
SMILES C(#N)C(Cl)(Cl)Cl

Specifications

Melting Point -42.0°C
Color Colorless to Yellow
Density 1.4400g/mL
Boiling Point 83.0°C to 84.0°C
Flash Point 195°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Linear Formula CCl3CN
Quantity 5 g
Beilstein 02,212
Fieser 01,1194; 05,686; 06,604; 07,381; 12,526; 13,32
Specific Gravity 1.44
Merck Index 15,9793
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Formula Weight 144.39
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trichloroacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 208-885-7

RUO – Research Use Only