missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetonitrile, 98%

Catalog No. AAA1056518
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1056518 50 g
AAA1056530 250 g
AAA105650B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1056518 Supplier Thermo Scientific Chemicals Supplier No. A1056518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 545-06-2 | C2Cl3N | 144.379 g/mol

Trichloroacetonitrile is involved as a reagent in Overman rearrangement, which is used to prepare alylic amines from allylic alcohols. It is also used to prepare bistrichloroacetimidates from diols leading to dihyrooxazines through acid catalyzed cyclization. Further, it is utilized in the synthesis of trichloroacetimidates by 1,8-Diazobicyclo[5.4.0]undec-7-ene (DBU) catalyzed addition of allylic alcohols. It finds application in the study of the methoxy methyl (MOM) catalyzed aza-Claisen rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetonitrile is involved as a reagent in Overman rearrangement, which is used to prepare alylic amines from allylic alcohols. It is also used to prepare bistrichloroacetimidates from diols leading to dihyrooxazines through acid catalyzed cyclization. Further, it is utilized in the synthesis of trichloroacetimidates by 1,8-Diazobicyclo[5.4.0]undec-7-ene (DBU) catalyzed addition of allylic alcohols. It finds application in the study of the methoxy methyl (MOM) catalyzed aza-Claisen rearrangement.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 545-06-2
Molecular Formula C2Cl3N
Molecular Weight (g/mol) 144.379
MDL Number MFCD00001842
InChI Key DRUIESSIVFYOMK-UHFFFAOYSA-N
Synonym trichloroacetonitrile, acetonitrile, trichloro, tritox, cyanotrichloromethane, trichloromethyl cyanide, trichloroethanenitrile, trichlouracetonitril, trichlor-acetonitrile, trichloromethylnitrile, nitrile trichloracetique
PubChem CID 11011
ChEBI CHEBI:82541
IUPAC Name 2,2,2-trichloroacetonitrile
SMILES C(#N)C(Cl)(Cl)Cl

Specifications

Melting Point -44°C
Density 1.44
Boiling Point 85°C to 86°C
Assay Percent Range 98%
Refractive Index 1.441
Linear Formula Cl3CCN
Quantity 50 g
UN Number UN3276
Beilstein 605572
Merck Index 14,9628
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 144.39
Percent Purity 98%
Chemical Name or Material Trichloroacetonitrile
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 208-885-7
RTECSNumber AM2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only