missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid 99.0+%, TCI America™
$59.65 - $192.00
Chemical Identifiers
| CAS | 76-03-9 |
|---|---|
| Molecular Formula | C2HCl3O2 |
| Molecular Weight (g/mol) | 163.378 |
| MDL Number | MFCD00004177 |
| InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
| Synonym | trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer |
| PubChem CID | 6421 |
| ChEBI | CHEBI:30956 |
| IUPAC Name | 2,2,2-trichloroacetic acid |
| SMILES | C(=O)(C(Cl)(Cl)Cl)O |
Chemical Identifiers
| 76-03-9 | |
| 163.378 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 6421 | |
| 2,2,2-trichloroacetic acid |
| C2HCl3O2 | |
| MFCD00004177 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| CHEBI:30956 | |
| C(=O)(C(Cl)(Cl)Cl)O |
Specifications
| 1.2 (0.1mol/L aq) | |
| 76-03-9 | |
| White | |
| MFCD00004177 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| C(=O)(C(Cl)(Cl)Cl)O | |
| 163.378 | |
| CHEBI:30956 | |
| ≥99.0% (T) | |
| Crystalline Powder |
| 58°C | |
| 198°C | |
| C2HCl3O2 | |
| 1839 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 2,2,2-trichloroacetic acid | |
| 6421 | |
| 163.38 | |
| 25 g | |
| Trichloroacetic Acid |
Safety and Handling
EINECSNumber : (2)-1188
RTECSNumber : AJ7875000
TSCA : Yes
Recommended Storage : Refrigerator