missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid 99.0+%, TCI America™
Specifications
Specifications
| pH | 1.2 (0.1mol/L aq) |
| Melting Point | 58°C |
| Boiling Point | 198°C |
| Color | White |
| Quantity | 500 g |
| Physical Form | Crystalline Powder |
| UN Number | 1839 |
| Formula Weight | 163.38 |
| Percent Purity | ≥99.0% (T) |
| Chemical Name or Material | Trichloroacetic Acid |
Safety and Handling
| EINECSNumber | (2)-1188 |
| RTECSNumber | AJ7875000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |