missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid 99.0+%, TCI America™

Catalog No. T036925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T036925G 25 g
T0369500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T036925G Supplier TCI America Supplier No. T036925G

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

pH 1.2 (0.1mol/L aq)
Melting Point 58°C
Boiling Point 198°C
Color White
UN Number 1839
Formula Weight 163.38
Percent Purity ≥99.0% (T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Trichloroacetic Acid
EINECSNumber (2)-1188
RTECSNumber AJ7875000
TSCA Yes
Recommended Storage Refrigerator