missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid. EMSURE™, MilliporeSigma™, For Analysis,

Catalog No. M1008070100
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1008070100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1008070100 Supplier Technidata Supplier No. 1.00807.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

pH <1 (50g/L, H2O, 20°C)
Melting Point 54°C to 56°C
Color Colorless
Quantity 100 g
Packaging Glass Bottle
Physical Form Deliquescent Crystals
Vapor Pressure 1 hPa (20 °C)
Solubility Information 1300g/L
Formula Weight 163.38
Percent Purity >99.5%
Grade ACS
Chemical Name or Material Trichloroacetic acid
Show More Show Less
EINECSNumber 200-927-2
RTECSNumber AJ7875000