missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid (Crystalline/Certified), Fisher Chemical™

Catalog No. p-202586
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A324500 500 g Glass Bottle
A3243 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A324500 Supplier Thermo Fisher Scientific Supplier No. FLA324500
Only null left

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

pH 1.2
Melting Point 52°C
Boiling Point 196°C
Color White
Assay Percent Range ≥99.0 %
Linear Formula CCl3COOH
UN Number 1839
Formula Weight 163.39
Percent Purity ≥99.0%
Grade Certified
Density 1.620g/cm³
Quantity 500 g
Ignition Residue 0.05% max.
Packaging Glass Bottle
Physical Form Solid
Vapor Pressure 1.2mbar at 50°C, 0.08mbar at 25°C
Chemical Name or Material Trichloroacetic Acid
Show More Show Less
DOTInformation DOT Class 8, : Corrosive