missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid (Crystalline/Certified), Fisher Chemical™
Specifications
Specifications
| pH | 1.2 |
| Melting Point | 52°C |
| Boiling Point | 196°C |
| Color | White |
| Density | 1.620g/cm³ |
| Quantity | 500 g |
| Ignition Residue | 0.05% max. |
| Packaging | Glass Bottle |
| Physical Form | Solid |
| Assay Percent Range | ≥99.0 % |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 8, : Corrosive |