missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, ACS, 99% min

Catalog No. p-7049756
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Catalog No. Quantity
AA2215618 50 g
AA2215630 250 g
AA22156A1 1 kg
3 options

Catalog No. AA2215618

Supplier: Thermo Scientific Chemicals 02215618

Only null left

Protein precipitation reagent | CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol

Protein precipitation reagentTrichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used in cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and as a decalcifier and fixative in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Melting Point 54°C to 58°C
Boiling Point 196°C
Assay Percent Range 99%
UN Number UN1839
Beilstein 970119
Merck Index 14,9627
Solubility Information Soluble in water,alcohol,ethyl ether,benzene,acetone,o-xylene and carbon tetrachloride.
Formula Weight 163.39
Percent Purity 99%
Grade ACS
Density 1.629
Quantity 50 g
Physical Form Crystalline
Sensitivity Hygroscopic
Chemical Name or Material Trichloroacetic acid, For ACS analysis
Show More Show Less
Hazard Category H314-H335-H351
Hazard Statement GHS H Statement
H314
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRICHLOROACETIC ACID
EINECSNumber 200-927-2
RTECSNumber AJ7875000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only