missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, 99%, extra pure

Catalog No. p-4112151
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC152135000 500 g Plastic bottle
AC152130010 1 kg Plastic bottle
2 options

Catalog No. AC152135000

Supplier: Thermo Scientific Chemicals 152135000

Only null left

CAS: 76-03-9 | C2HCl3O2 | 163.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.39
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Melting Point 52.0°C to 58.0°C
Boiling Point 196.0°C
Color White
Assay Percent Range 0.99
Linear Formula CCl3CO2H
Beilstein 2206
Fieser 01,1194; 02,425; 04,520; 15,168
Merck Index 159792
Solubility Information Solubility in water: 120g/100 mL (20°C). Other solubilities: soluble in alcohol and ether,carbon tetrachloride
Formula Weight 163.39
Percent Purity 99%
Grade Extra Pure
Density 1.6200 g/mL
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Flakes, Crystals, or Crystalline Powder
Specific Gravity 1.62
Chemical Name or Material Trichloroacetic acid, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 200-927-2

RUO – Research Use Only