missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, 99+%, ACS reagent

Catalog No. AC421451000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421451000 100 g Glass Bottle
AC421455000 500 g Glass Bottle
AC421450025 2.5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC421451000 Supplier Thermo Scientific Chemicals Supplier No. 421451000

CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Color White
Assay Percent Range 99+%
Linear Formula CCl3CO2H
Beilstein 02, 206
Fieser 01,1194; 02,425; 04,520; 15,168
Merck Index 15, 9792
Formula Weight 163.39
Percent Purity ≥99.0%
Grade ACS Reagent
Density 1.6200g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Adhering Crystals
Residue after Ignition 0.03% max.
Vapor Pressure 1.2mbar at 50°C
Chemical Name or Material Trichloroacetic acid, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-927-2
RTECSNumber AJ7875000
TSCA TSCA

RUO – Research Use Only