missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, 5% w/v aq. soln.

Catalog No. AAJ62366K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62366K2 1 L
AAJ62366AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62366K2 Supplier Thermo Scientific Chemicals Supplier No. J62366K2

missing translation for 'validMainframeMsg'

Protein precipitation reagent | CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol

Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

Solubility
Soluble in water (1200 mg/ml at 20°C), alcohol, ether, ethanol (very soluble), and carbon tetrachloride.

Notes
Store under nitrogen. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases, smines.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Color Colorless
Concentration 5% w/v aq. soln.
Quantity 1 L
Odor Acrid
Physical Form Liquid
Linear Formula Cl3CCO2H
UN Number UN2564
Beilstein 970119
Merck Index 14,9627
Solubility Information Soluble in water (1200mg/ml at 20°C),alcohol,ether,ethanol (very soluble),and carbon tetrachloride.
Formula Weight 163.39
Chemical Name or Material Trichloroacetic acid
Show More Show Less
Hazard Category H314-H335-H351
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: TRICHLOROACETIC ACID, SOLUTION
EINECSNumber 200-927-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only