Learn More
Trichloroacetic acid, 5% w/v aq. soln.
Description
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.
Solubility
Soluble in water (1200 mg/ml at 20°C), alcohol, ether, ethanol (very soluble), and carbon tetrachloride.
Notes
Store under nitrogen. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases, smines.
Specifications
Specifications
| Color | Colorless |
| Concentration | 5% w/v aq. soln. |
| Quantity | 500 mL |
| Odor | Acrid |
| Physical Form | Liquid |
| Linear Formula | Cl3CCO2H |
| UN Number | UN2564 |
| Beilstein | 970119 |
| Merck Index | 14,9627 |
| Solubility Information | Soluble in water (1200mg/ml at 20°C),alcohol,ether,ethanol (very soluble),and carbon tetrachloride. |
| Show More |
Safety and Handling
| Hazard Category | H314-H335-H351 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: TRICHLOROACETIC ACID, SOLUTION |
| EINECSNumber | 200-927-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only