missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 33% (w/v), Ricca Chemical

Catalog No. 868831
Change view
Click to view available options
Package: Amber Glass; Size: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
868831 Package: Amber Glass; Size: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 868831 Supplier Ricca Chemical Company Supplier No. 8688.31

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Specifications

CAS 76-03-9, 7732-18-5, 7440-44-0
CAS Min % 24.29, 74.0, 0.15
CAS Max % 25.28, 77.0, 0.15
Molecular Formula C2HCl3O2
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES OC(=O)C(Cl)(Cl)Cl
IUPAC Name trichloroacetic acid
Molecular Weight (g/mol) 163.38
PubChem CID 6421
ChEBI CHEBI:30956
Concentration or Composition (by Analyte or Components) 33% (w/v) Aqueous Solution
Quantity 4 L
Packaging Amber Glass Bottle
Chemical Name or Material Trichloroacetic Acid
Show More Show Less
ShelfLife 12 months