missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 30% (w/v) Aqueous Solution, Spectrum™ Chemical

$447.35 - $600.50

Chemical Identifiers

CAS 76-03-9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614510
View Documents
Spectrum Chemical Mfg Cor
T181500MLGL
500 mL Amber Glass Bottle
Each for $447.35
Only null left
 
18614511
View Documents
Spectrum Chemical Mfg Cor
T1811LTGL
1 L Amber Glass Bottle
Each for $600.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

76-03-9

Specifications

76-03-9,7732-18-5
Chemical Solution
Amber Glass Bottle
30% ±1.2%
500 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.