missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 20% (w/v), Ricca Chemical

Catalog No. 868732
Change view
Click to view available options
Package: Amber Glass; Size: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
868732 Package: Amber Glass; Size: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 868732 Supplier Ricca Chemical Company Supplier No. 868732

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 76-03-9, 7732-18-5
Molecular Formula C2HCl3O2
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES C(=O)(C(Cl)(Cl)Cl)O
IUPAC Name 2,2,2-trichloroacetic acid
Molecular Weight (g/mol) 163.378
PubChem CID 6421
ChEBI CHEBI:30956
Quantity 1 L
Packaging Amber Glass Bottle
Chemical Name or Material Trichloroacetic acid
Show More Show Less