missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetamide 98.0+%, TCI America™

Catalog No. T0368500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0368500G 500 g
T036825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0368500G Supplier TCI America Supplier No. T0368500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 594-65-0
Molecular Formula C2H2Cl3NO
Molecular Weight (g/mol) 162.39
MDL Number MFCD00008009
InChI Key UPQQXPKAYZYUKO-UHFFFAOYSA-N
Synonym trichloroacetamide, acetamide, 2,2,2-trichloro, amid kyseliny trichloroctove, acetamide, alpha-trichloro, unii-i8bl3305ro, amid kyseliny trichloroctove czech, 2,2,2-trichloro-acetamide, acetamide, .alpha.-trichloro, .alpha.,.alpha.,.alpha.-trichloroacetamide, trichloro-acetamide
PubChem CID 61144
IUPAC Name trichloroacetamide
SMILES NC(=O)C(Cl)(Cl)Cl

Specifications

Melting Point 141°C
Color White
Boiling Point 240°C
Quantity 500 g
Formula Weight 162.39
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Trichloroacetamide
RTECSNumber AC9275000
TSCA Yes