missing translation for 'onlineSavingsMsg'
Learn More

Trichloro-2-cyanoethylsilane 98.0+%, TCI America™

Catalog No. T132225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T132225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T132225G Supplier TCI America Supplier No. T132225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1071-22-3
Molecular Formula C3H4Cl3NSi
Molecular Weight (g/mol) 188.507
MDL Number MFCD00039493
InChI Key OLBGECWYBGXCNV-UHFFFAOYSA-N
Synonym 2-Cyanoethyltrichlorosilane, Trichlorosilylpropionitrile
PubChem CID 14061
IUPAC Name 3-trichlorosilylpropanenitrile
SMILES C(C[Si](Cl)(Cl)Cl)C#N

Specifications

Color White-Yellow
Boiling Point 109°C
Quantity 25 g
UN Number 2987
Formula Weight 188.51
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Trichloro-2-cyanoethylsilane
RTECSNumber UG4900000
TSCA Yes