missing translation for 'onlineSavingsMsg'
Learn More

Trichloro(1H,1H,2H,2H-tridecafluoro-n-octyl)silane 97.0+%, TCI America™

Catalog No. p-7137292
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25775G 5 g
T257725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25775G Supplier TCI America Supplier No. T25775G
Only null left

Chemical Identifiers

CAS 78560-45-9
Molecular Formula C8H4Cl3F13Si
Molecular Weight (g/mol) 481.534
MDL Number MFCD00042363
InChI Key PISDRBMXQBSCIP-UHFFFAOYSA-N
Synonym trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane
PubChem CID 123578
IUPAC Name trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 85°C
Quantity 5 g
UN Number 2986
Formula Weight 481.53
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichloro(1H,1H,2H,2H-tridecafluoro-n-octyl)silane
EINECSNumber (2)-2046
TSCA No