missing translation for 'onlineSavingsMsg'
Learn More

Tricarbonylnitrosylcobalt

Catalog No. p-7049822
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2315103 1 g
AA2315106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2315103 Supplier Thermo Scientific Chemicals Supplier No. 02315103
Only null left

CAS: 14096-82-3

Tricarbonylnitrosylcobalt is used as the Co metalorganic precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14096-82-3
MDL Number MFCD00016014
Synonym cobalt tricarbonyl nitrosyl, tricarbonyl-nitrosyl-cobalt

Specifications

Melting Point -1°C
Color Red
Density 1.47 g/mL
Boiling Point 48.6°C (55°C (decomposition))
Odor Unpleasant
Linear Formula Co(NO)(CO)3
Quantity 1 g
UN Number UN1992
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 172.97
Physical Form Liquid
Chemical Name or Material Tricarbonylnitrosylcobalt
Show More Show Less
Hazard Category H225-H301-H330-H334-H350
Hazard Statement GHS H Statement
H225-H301-H330-H351-H317
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P284-P285-P301+P310-P303+P361+P353-P304+P340-P308+P313-P310-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 237-945-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only