missing translation for 'onlineSavingsMsg'
Learn More

Tricarballylic acid, 98%

Catalog No. AAB2210306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2210306 5 g
AAB2210314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2210306 Supplier Thermo Scientific Chemicals Supplier No. B2210306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-14-9 | C6H8O6 | 176.12 g/mol

It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

Solubility
Soluble in water (500 mg/ml at 18°C), methanol, and ether (slightly).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 99-14-9
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00002723
InChI Key KQTIIICEAUMSDG-UHFFFAOYSA-N
Synonym tricarballylic acid, 1,2,3-propanetricarboxylic acid, carballylic acid, tricarballylate, beta-carboxyglutaric acid, propane 1,2,3-tricarboxylic acid, unii-ra5qh2j020, 3-carboxyglutaric acid, .beta.-carboxyglutaric acid, 1,2,3-propanetricarboxylicacid
PubChem CID 14925
ChEBI CHEBI:45969
IUPAC Name propane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(CC(O)=O)C(O)=O

Specifications

Melting Point 157°C to 162°C
Quantity 5 g
Beilstein 1783567
Merck Index 14,9622
Solubility Information Soluble in water (500mg/ml at 18°C),methanol,and ether (slightly).
Formula Weight 176.12
Percent Purity 98%
Chemical Name or Material Tricarballylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-733-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only