missing translation for 'onlineSavingsMsg'
Learn More

Tricaproin 95.0+%, TCI America™

Catalog No. T044110ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T044110ML 10 mL
T044125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T044110ML Supplier TCI America Supplier No. T044110ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-70-5
Molecular Formula C21H38O6
Molecular Weight (g/mol) 386.529
MDL Number MFCD00042905
InChI Key MAYCICSNZYXLHB-UHFFFAOYSA-N
Synonym Glycerol Trihexanoate, Trihexanoin
PubChem CID 12131
ChEBI CHEBI:77386
IUPAC Name 2,3-di(hexanoyloxy)propyl hexanoate
SMILES CCCCCC(=O)OCC(COC(=O)CCCCC)OC(=O)CCCCC

Specifications

Color Yellow
Boiling Point 244°C
Quantity 10 mL
Formula Weight 386.53
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tricaproin
EINECSNumber (2)-0666
RTECSNumber MB2700000
TSCA Yes