missing translation for 'onlineSavingsMsg'
Learn More

Tricaprin 98.0+%, TCI America™

Catalog No. p-7136267
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T041310G 10 g
T041325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T041310G Supplier TCI America Supplier No. T041310G

Chemical Identifiers

CAS 621-71-6
Molecular Formula C33H62O6
Molecular Weight (g/mol) 554.853
MDL Number MFCD00036239
InChI Key LADGBHLMCUINGV-UHFFFAOYSA-N
Synonym tricaprin, tridecanoin, glycerol tricaprate, glycerol tridecanoate, decanoin, tri, tricapric glyceride, glycerol tricaprin, 1,2,3-tridecanoylglycerol, capric acid triglyceride, glyceryl tricaprate
PubChem CID 69310
ChEBI CHEBI:77388
IUPAC Name 2,3-di(decanoyloxy)propyl decanoate
SMILES CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC

Specifications

Melting Point 32°C
Color White
Boiling Point 254°C
Quantity 10 g
Formula Weight 554.85
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tricaprin
EINECSNumber (2)-0669
RTECSNumber HE0100000
TSCA Yes
Recommended Storage Refrigerator