missing translation for 'onlineSavingsMsg'
Learn More

Tricaprin, Approx. 99%, MP Biomedicals™

Catalog No. ICN10529705
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10529705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10529705 Supplier MP Biomedicals Inc Supplier No. 0210529705

Chemical Identifiers

CAS 621-71-6
Molecular Formula C33H62O6
Molecular Weight (g/mol) 554.85
InChI Key LADGBHLMCUINGV-UHFFFAOYSA-N
Synonym tricaprin, tridecanoin, glycerol tricaprate, glycerol tridecanoate, decanoin, tri, tricapric glyceride, glycerol tricaprin, 1,2,3-tridecanoylglycerol, capric acid triglyceride, glyceryl tricaprate
PubChem CID 69310
ChEBI CHEBI:77388
IUPAC Name 1,3-bis(decanoyloxy)propan-2-yl decanoate
SMILES CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC

Specifications

Assay Percent Range approx.99%
Quantity 5 g
Formula Weight 554.8
Physical Form Solid
Chemical Name or Material Tricaprin
Recommended Storage Store at 0°C.