missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyrin, 98.4%, MP Biomedicals™
$120.50
Chemical Identifiers
| CAS | 60-01-5 |
|---|---|
| Molecular Formula | C15H26O6 |
| InChI Key | UYXTWWCETRIEDR-UHFFFAOYSA-N |
| SMILES | CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10311180
|
N/A | 100 mL |
Each for $120.50
|
|
|||||
Chemical Identifiers
| 60-01-5 | |
| C15H26O6 | |
| UYXTWWCETRIEDR-UHFFFAOYSA-N | |
| CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Specifications
| 60-01-5 | |
| 1.032g/mL | |
| C15H26O6 | |
| UYXTWWCETRIEDR-UHFFFAOYSA-N | |
| 2,3-di(butanoyloxy)propyl butanoate | |
| 6050 | |
| 302.36 | |
| Tributyrin |
| Colorless | |
| 0.984 | |
| tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
| CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
| 302.367 | |
| CHEBI:35020 | |
| Liquid |