missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylphosphine Oxide 95.0+%, TCI America™
Supplier: TCI America T092125G
Specifications
| Tributylphosphine Oxide | |
| 71°C | |
| C12H27OP | |
| 25 g | |
| tributylphosphine oxide, tri-n-butylphosphine oxide, butyphos, phosphine oxide, tributyl, trisbutylphosphine oxide, tributylfosfinoxid, tributylfosfinoxid czech, unii-w58cdf8s4h, tributylphosphane oxide, w58cdf8s4h | |
| CCCCP(=O)(CCCC)CCCC | |
| 218.321 | |
| 218.32 | |
| Crystalline Powder |
| 814-29-9 | |
| White-Yellow | |
| MFCD00002082 | |
| 1759 | |
| MNZAKDODWSQONA-UHFFFAOYSA-N | |
| 1-dibutylphosphorylbutane | |
| 13138 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 814-29-9 | |
| 218.321 | |
| MNZAKDODWSQONA-UHFFFAOYSA-N | |
| 13138 | |
| CCCCP(=O)(CCCC)CCCC |
| C12H27OP | |
| MFCD00002082 | |
| tributylphosphine oxide, tri-n-butylphosphine oxide, butyphos, phosphine oxide, tributyl, trisbutylphosphine oxide, tributylfosfinoxid, tributylfosfinoxid czech, unii-w58cdf8s4h, tributylphosphane oxide, w58cdf8s4h | |
| 1-dibutylphosphorylbutane |
Safety and Handling
RTECSNumber : SZ1575000
TSCA : Yes