missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylmethylammonium Chloride (ca. 75% in water), TCI America™
Supplier: TCI America T2873100ML
Specifications
| Tributylmethylammonium Chloride (ca. 75% in water) | |
| Yellow | |
| MFCD00011847 | |
| tributylmethylammonium chloride, n,n-dibutyl-n-methylbutan-1-aminium chloride, methyltributylammonium chloride, methyl tributyl ammonium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride, unii-az0b34me3u, az0b34me3u, tributyl methyl azanium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride 1:1, acmc-20aker | |
| CCCC[N+](C)(CCCC)CCCC.[Cl-] | |
| 235.84 | |
| 235.84 |
| 56375-79-2 | |
| C13H30ClN | |
| 100 mL | |
| IPILPUZVTYHGIL-UHFFFAOYSA-M | |
| tributyl(methyl)azanium;chloride | |
| 91822 | |
| Liquid |
Chemical Identifiers
| 56375-79-2 | |
| 235.84 | |
| IPILPUZVTYHGIL-UHFFFAOYSA-M | |
| 91822 | |
| CCCC[N+](C)(CCCC)CCCC.[Cl-] |
| C13H30ClN | |
| MFCD00011847 | |
| tributylmethylammonium chloride, n,n-dibutyl-n-methylbutan-1-aminium chloride, methyltributylammonium chloride, methyl tributyl ammonium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride, unii-az0b34me3u, az0b34me3u, tributyl methyl azanium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride 1:1, acmc-20aker | |
| tributyl(methyl)azanium;chloride |
Safety and Handling
TSCA : Yes