missing translation for 'onlineSavingsMsg'
Learn More

Tributylhexadecylphosphonium Bromide 98.0+%, TCI America™

Catalog No. H104725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H104725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H104725G Supplier TCI America Supplier No. H104725G

Chemical Identifiers

CAS 14937-45-2
Molecular Formula C28H60BrP
Molecular Weight (g/mol) 507.67
MDL Number MFCD00011775
InChI Key RYVBINGWVJJDPU-UHFFFAOYSA-M
Synonym tributylhexadecylphosphonium bromide, tributyl hexadecyl phosphonium bromide, cetyltributylphosphonium bromide, tributyl hexadecyl phosphanium bromide, phosphonium, tributylhexadecyl-, bromide, hexadecyltributylphosphonium bromide, phosphonium, tributylhexadecyl-, bromide 1:1, hexadecyltri-n-butylphosphonium bromide, n-hexadecyl tri-n-butylphosphonium bromide, tbhdpb
PubChem CID 84716
IUPAC Name tributyl(hexadecyl)phosphanium bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC

Specifications

Melting Point 61°C
Color White
Quantity 25 g
Formula Weight 507.67
Percent Purity ≥98.0% (T)
Physical Form Crystals
Chemical Name or Material Tributylhexadecylphosphonium Bromide