missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyldodecylphosphonium Bromide 98.0+%, TCI America™
Supplier: TCI America D291225G
Specifications
| Tributyldodecylphosphonium Bromide | |
| White-Yellow | |
| MFCD04038166 | |
| Dodecyltributylphosphonium Bromide, Lauryltributylphosphonium Bromide | |
| CCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC.[Br-] | |
| 451.558 | |
| 451.56 | |
| Crystalline Lumps |
| 15294-63-0 | |
| C24H52BrP | |
| 25 g | |
| ABJGUZZWSKMTEI-UHFFFAOYSA-M | |
| tributyl(dodecyl)phosphanium;bromide | |
| 11730092 | |
| ≥98.0% (T) |
Chemical Identifiers
| 15294-63-0 | |
| 451.558 | |
| ABJGUZZWSKMTEI-UHFFFAOYSA-M | |
| 11730092 | |
| CCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC.[Br-] |
| C24H52BrP | |
| MFCD04038166 | |
| Dodecyltributylphosphonium Bromide, Lauryltributylphosphonium Bromide | |
| tributyl(dodecyl)phosphanium;bromide |
Safety and Handling
TSCA : No