missing translation for 'onlineSavingsMsg'
Learn More

Tributylchlorosilane 97.0+%, TCI America™

Catalog No. p-7137783
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35245G 5 g
T352425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35245G Supplier TCI America Supplier No. T35245G
Only null left

Chemical Identifiers

CAS 995-45-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00000509
InChI Key JSQJUDVTRRCSRU-UHFFFAOYSA-N
Synonym silane, tributylchloro, chlorotributylsilane, chlorotri-n-butylsilane, tributyl chloro silane, tri-n-butylchlorosilane, tributylsilyl chloride, tributylchloro-silan, chloro-tri-n-butylsilane
PubChem CID 70448
IUPAC Name tributyl(chloro)silane
SMILES CCCC[Si](CCCC)(CCCC)Cl

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
UN Number 2987
Formula Weight 234.88
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributylchlorosilane
EINECSNumber (2)-2041
TSCA Yes