missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl Phosphite 93.0+%, TCI America™
$67.11 - $251.65
Chemical Identifiers
| CAS | 102-85-2 |
|---|---|
| Molecular Formula | C12H27O3P |
| Molecular Weight (g/mol) | 250.319 |
| MDL Number | MFCD00009437 |
| InChI Key | XTTGYFREQJCEML-UHFFFAOYSA-N |
| Synonym | tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri |
| PubChem CID | 7623 |
| IUPAC Name | tributyl phosphite |
| SMILES | CCCCOP(OCCCC)OCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T036225ML
|
TCI America
T036225ML |
25 mL |
Each for $67.11
|
|
|||||
|
T0362500ML
|
TCI America
T0362500ML |
500 mL |
Each for $251.65
|
|
|||||
Chemical Identifiers
| 102-85-2 | |
| 250.319 | |
| XTTGYFREQJCEML-UHFFFAOYSA-N | |
| 7623 | |
| CCCCOP(OCCCC)OCCCC |
| C12H27O3P | |
| MFCD00009437 | |
| tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri | |
| tributyl phosphite |
Specifications
| 102-85-2 | |
| Yellow | |
| C12H27O3P | |
| 25 mL | |
| XTTGYFREQJCEML-UHFFFAOYSA-N | |
| tributyl phosphite | |
| 7623 | |
| ≥95.0% (GC) | |
| Tributyl Phosphite |
| -80°C | |
| 125°C | |
| MFCD00009437 | |
| tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri | |
| CCCCOP(OCCCC)OCCCC | |
| 250.319 | |
| 250.32 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2906
RTECSNumber : TH0875000
TSCA : Yes