missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl phosphate, For extraction analysis (GC), ≥99.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | -79°C |
| CAS | 126-73-8 |
| Density | 0.979 g/mL (at 25°C) |
| Boiling Point | 180°C to 183°C (22 mmHg) |
| Flash Point | 145°C |
| Ignition Point | 770°F |
| Linear Formula | (CH3(CH2)3 O)3 PO |
| Quantity | 500 mL |
| Vapor Density | 9.2 (vs. air) |
| Formula Weight | 266.31 |
| Show More |
Safety and Handling
| RTECSNumber | TC7700000 |