missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl O-Acetylcitrate 97.0+%, TCI America™
$31.68 - $100.67
Chemical Identifiers
| CAS | 77-90-7 |
|---|---|
| Molecular Formula | C20H34O8 |
| Molecular Weight (g/mol) | 402.48 |
| MDL Number | MFCD00043554 |
| InChI Key | QZCLKYGREBVARF-UHFFFAOYSA-N |
| Synonym | O-Acetylcitric Acid Tributyl Ester |
| PubChem CID | 6505 |
| IUPAC Name | 1,2,3-tributyl 2-(acetyloxy)propane-1,2,3-tricarboxylate |
| SMILES | CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A082225ML
|
TCI America
A082225ML |
25 mL |
Each for $31.68
|
|
|||||
|
A0822500ML
|
TCI America
A0822500ML |
500 mL |
Each for $100.67
|
|
|||||
Chemical Identifiers
| 77-90-7 | |
| 402.48 | |
| QZCLKYGREBVARF-UHFFFAOYSA-N | |
| 6505 | |
| CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC |
| C20H34O8 | |
| MFCD00043554 | |
| O-Acetylcitric Acid Tributyl Ester | |
| 1,2,3-tributyl 2-(acetyloxy)propane-1,2,3-tricarboxylate |
Specifications
| 77-90-7 | |
| Yellow | |
| C20H34O8 | |
| 25 mL | |
| QZCLKYGREBVARF-UHFFFAOYSA-N | |
| 1,2,3-tributyl 2-(acetyloxy)propane-1,2,3-tricarboxylate | |
| 6505 | |
| ≥97.0% (GC) | |
| Tributyl O-Acetylcitrate |
| -80°C | |
| 343°C | |
| MFCD00043554 | |
| O-Acetylcitric Acid Tributyl Ester | |
| CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC | |
| 402.48 | |
| 402.48 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1327
RTECSNumber : TZ8330000
TSCA : Yes