missing translation for 'onlineSavingsMsg'
Learn More

Tributyl-n-octylphosphonium Bromide 98.0+%, TCI America™

Catalog No. O029725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O029725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O029725G Supplier TCI America Supplier No. O029725G
Only null left

Chemical Identifiers

CAS 57702-65-5
Molecular Formula C20H44BrP
Molecular Weight (g/mol) 395.45
MDL Number MFCD04038147
InChI Key UJMLRSWRUXXZEW-UHFFFAOYSA-M
Synonym n-Octyltributylphosphonium Bromide
PubChem CID 22667357
IUPAC Name tributyl(octyl)phosphanium;bromide
SMILES CCCCCCCC[P+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 395.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Tributyl-n-octylphosphonium Bromide