missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(cyanomethyl)phosphonium Chloride 98.0+%, TCI America™

Catalog No. C144210G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C144210G 10 g
C144225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C144210G Supplier TCI America Supplier No. C144210G
Only null left

Chemical Identifiers

CAS 82358-61-0
Molecular Formula C14H29ClNP
Molecular Weight (g/mol) 277.817
MDL Number MFCD00671508
InChI Key JCHWNYYARSRCKZ-UHFFFAOYSA-M
Synonym (Cyanomethyl)tributylphosphonium Chloride
PubChem CID 11594300
IUPAC Name tributyl(cyanomethyl)phosphanium;chloride
SMILES CCCC[P+](CCCC)(CCCC)CC#N.[Cl-]

Specifications

Melting Point 97°C
Color White
Quantity 10 g
UN Number 3439
Formula Weight 277.82
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tributyl(cyanomethyl)phosphonium Chloride