missing translation for 'onlineSavingsMsg'
Learn More

Tributyl Borate 99.0+%, TCI America™

Catalog No. B051825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B051825ML 25 mL
B0518100ML 100 mL
B0518500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B051825ML Supplier TCI America Supplier No. B051825ML

Chemical Identifiers

CAS 688-74-4
Molecular Formula C12H27BO3
Molecular Weight (g/mol) 230.155
MDL Number MFCD00009434
InChI Key LGQXXHMEBUOXRP-UHFFFAOYSA-N
Synonym n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
PubChem CID 12712
IUPAC Name tributyl borate
SMILES B(OCCCC)(OCCCC)OCCCC

Specifications

Color Colorless
Boiling Point 230°C
Quantity 25 mL
Formula Weight 230.16
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Tributyl Borate
EINECSNumber (2)-2101
RTECSNumber ED4900000
TSCA Yes