Learn More
Tributyl borate, 98%, AcroSeal™
CAS: 688-74-4 | C12H27BO3 | 230.15 g/mol
$110.61 - $110.61
Chemical Identifiers
| CAS | 688-74-4 |
|---|---|
| Molecular Formula | C12H27BO3 |
| Molecular Weight (g/mol) | 230.15 |
| MDL Number | MFCD00009434 |
| InChI Key | LGQXXHMEBUOXRP-UHFFFAOYSA-N |
| Synonym | n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy |
| PubChem CID | 12712 |
| IUPAC Name | tributyl borate |
| SMILES | B(OCCCC)(OCCCC)OCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC428661000
|
Thermo Scientific Chemicals
428661000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $110.61
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 688-74-4 | |
| 230.15 | |
| LGQXXHMEBUOXRP-UHFFFAOYSA-N | |
| 12712 | |
| B(OCCCC)(OCCCC)OCCCC |
| C12H27BO3 | |
| MFCD00009434 | |
| n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy | |
| tributyl borate |
Specifications
| 688-74-4 | |
| Colorless | |
| 230.0°C to 235.0°C | |
| Authentic | |
| AcroSeal™ Glass Bottle | |
| 1.4080 to 1.4100 | |
| MFCD00009434 | |
| 0.853 | |
| Solubility in water: decomposes. Other solubilities: miscible with most common organic liquids | |
| B(OCCCC)(OCCCC)OCCCC | |
| 230.15 | |
| 230.15 | |
| Liquid |
| -70.0°C | |
| 0.8530g/mL | |
| 93°C | |
| 97.5% min. (GC) | |
| C12H27BO3 | |
| (CH3CH2CH2CH2O)3B | |
| 100 mL | |
| n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy | |
| LGQXXHMEBUOXRP-UHFFFAOYSA-N | |
| tributyl borate | |
| 12712 | |
| 98% | |
| Tributyl borate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 211-706-5
RUO – Research Use Only