missing translation for 'onlineSavingsMsg'
Learn More

tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane, Thermo Scientific™

Catalog No. CC58241DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC58241DA 1 g Amber Glass Bottle
CC58241CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC58241DA Supplier Thermo Scientific Chemicals Supplier No. CC58241DA

Chemical Identifiers

CAS 1007847-70-2
Molecular Formula C22H39NO2Sn
Molecular Weight (g/mol) 468.269
InChI Key TYKSDTUCXMRYNR-UHFFFAOYSA-N
Synonym 2-oxan-4-yloxy-6-tributylstannyl pyridine, tributyl 6-tetrahydropyran-4-yloxy pyrid-2-yl stannane, 2-oxan-4-yl oxy-6-tributylstannyl pyridine
PubChem CID 45594295
IUPAC Name tributyl-[6-(oxan-4-yloxy)pyridin-2-yl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)OC2CCOCC2

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 468.25
Percent Purity 90%
Chemical Name or Material tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane
Hazard Category GHS Signal Word: Danger