missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(1-ethoxyvinyl)tin, 95%

Catalog No. p-3737604
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377710010 1 g Glass bottle
AC377710050 5 g Glass bottle
AC377710250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC377710010 Supplier Thermo Scientific Chemicals Supplier No. 377710010
Only null left

CAS: 97674-02-7 | C16H34OSn | 361.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97674-02-7
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00010240
InChI Key HGXJOXHYPGNVNK-UHFFFAOYSA-N
Synonym tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl
PubChem CID 619414
IUPAC Name tributyl(1-ethoxyethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C(=C)OCC

Specifications

Color Colorless
Density 1.0690g/mL
Boiling Point 85.0°C to 86.0°C (0.1 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.4770
Linear Formula [CH3(CH2)3]SnC(CH2)OCH2CH3
Quantity 1 g
Specific Gravity 1.069
Solubility Information Solubility in water: insoluble
Formula Weight 361.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tributyl(1-ethoxyvinyl)tin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN

RUO – Research Use Only