missing translation for 'onlineSavingsMsg'
Learn More

Tribromofluoromethane 98.0+%, TCI America™

Catalog No. T23045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23045G Supplier TCI America Supplier No. T23045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 353-54-8
Molecular Formula CBr3F
Molecular Weight (g/mol) 270.721
MDL Number MFCD00000366
InChI Key IHZAEIHJPNTART-UHFFFAOYSA-N
Synonym fluorotribromomethane, methane, tribromofluoro, tribromo fluoro methane, tribromo-fluoro-methane, fluortribrommethan, tribrom fluor methan, cfbr3, fluoro tribromo methane, acmc-209igd, tribromofluoromethane
PubChem CID 67707
IUPAC Name tribromo(fluoro)methane
SMILES C(F)(Br)(Br)Br

Specifications

Melting Point -73°C
Color Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 270.72
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tribromofluoromethane