missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triarachidin 98.0+%, TCI America™
$106.25 - $533.13
Chemical Identifiers
| CAS | 620-64-4 |
|---|---|
| Molecular Formula | C63H122O6 |
| Molecular Weight (g/mol) | 975.663 |
| MDL Number | MFCD00042879 |
| InChI Key | KCVWRCXEUJUXIG-UHFFFAOYSA-N |
| Synonym | Glycerol Trieicosanoate, Triarachin, 1,2,3-Trieicosanoyl Glycerol |
| PubChem CID | 522017 |
| IUPAC Name | 2,3-di(icosanoyloxy)propyl icosanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 620-64-4 | |
| 975.663 | |
| KCVWRCXEUJUXIG-UHFFFAOYSA-N | |
| 522017 | |
| CCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCC |
| C63H122O6 | |
| MFCD00042879 | |
| Glycerol Trieicosanoate, Triarachin, 1,2,3-Trieicosanoyl Glycerol | |
| 2,3-di(icosanoyloxy)propyl icosanoate |
Specifications
| 620-64-4 | |
| White | |
| MFCD00042879 | |
| Glycerol Trieicosanoate, Triarachin, 1,2,3-Trieicosanoyl Glycerol | |
| CCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCC | |
| 975.663 | |
| 975.66 | |
| Crystalline Powder |
| 78°C | |
| C63H122O6 | |
| 1 g | |
| KCVWRCXEUJUXIG-UHFFFAOYSA-N | |
| 2,3-di(icosanoyloxy)propyl icosanoate | |
| 522017 | |
| ≥98.0% (GC) | |
| Triarachidin |
Safety and Handling
TSCA : No