Learn More
Triacetoxy(methyl)silane, tech. 90%
CAS: 4253-34-3 | C7H12O6Si | 220.25 g/mol
Supplier: Thermo Scientific Chemicals B2397418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 87°C to 88°C (3mmHg) | |
| 40°C to 45°C | |
| 50 g | |
| MFCD00008694 | |
| 1788668 | |
| methyltriacetoxysilane, methylsilanetriyl triacetate, triacetoxymethylsilane, triacetoxy methyl silane, silanetriol, methyl-, triacetate, silane, methyltriacetoxy, methylsilanetriol triacetate, methyltrihydroxysilane triacetate, apk 1 silane derivative, triacetoxy-methylsilane | |
| CC(=O)O[Si](C)(OC(C)=O)OC(C)=O | |
| 220.25 | |
| 220.25 | |
| Moisture sensitive |
| Triacetoxy(methyl)silane | |
| 4253-34-3 | |
| C7H12O6Si | |
| UN1759 | |
| 85°C (185°F) | |
| TVJPBVNWVPUZBM-UHFFFAOYSA-N | |
| [diacetyloxy(methyl)silyl] acetate | |
| 77929 | |
| 96% | |
| 1.175 |
Chemical Identifiers
| 4253-34-3 | |
| 220.25 | |
| TVJPBVNWVPUZBM-UHFFFAOYSA-N | |
| 77929 |
| C7H12O6Si | |
| MFCD00008694 | |
| methyltriacetoxysilane, methylsilanetriyl triacetate, triacetoxymethylsilane, triacetoxy methyl silane, silanetriol, methyl-, triacetate, silane, methyltriacetoxy, methylsilanetriol triacetate, methyltrihydroxysilane triacetate, apk 1 silane derivative, triacetoxy-methylsilane | |
| CC(=O)O[Si](C)(OC(C)=O)OC(C)=O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 224-221-9
RTECSNumber : VV4500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only