missing translation for 'onlineSavingsMsg'
Learn More

Triacetin, USP, 97-102%, Spectrum™ Chemical

Catalog No. p-7158526
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607505 500 mL Amber Glass Bottle
18607509 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18607505 Supplier Spectrum Chemical Mfg Cor Supplier No. TR106500MLGL
Only null left

TR106, 102-76-1, C9H14O6

Spectrum™ Chemical Triacetin, USP is used as an antifungal agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
IUPAC Name 1,3-bis(acetyloxy)propan-2-yl acetate
SMILES CC(=O)OCC(COC(C)=O)OC(C)=O

Specifications

CAS Max % 1
Boiling Point 258°C
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 97 to 102%
Grade USP
Physical Form Liquid
Chemical Name or Material Triacetin