missing translation for 'onlineSavingsMsg'
Learn More

Triacetin 99.0+%, TCI America™

Catalog No. p-7130136
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G008625G 25 g
G0086500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G008625G Supplier TCI America Supplier No. G008625G
Only null left

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.205
MDL Number MFCD00008716
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
Synonym triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
PubChem CID 5541
ChEBI CHEBI:9661
IUPAC Name 2,3-diacetyloxypropyl acetate
SMILES CC(=O)OCC(COC(=O)C)OC(=O)C

Specifications

Melting Point -78°C
Color Colorless
Boiling Point 260°C
Quantity 25 g
Formula Weight 218.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Triacetin
EINECSNumber (2)-0666&(2)-0753
RTECSNumber AK3675000
TSCA Yes