missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triacetin (Glycerol Triacetate), Reagents
Specifications
Specifications
| Boiling Range | 258°C to 259°C |
| Color | Colorless |
| Boiling Point | 258°C |
| Packaging | Glass Bottle |
| Quantity | 120 mL |
| Specific Gravity | 1.156 |
| Grade | Reagent |
| Physical Form | Liquid |
| Chemical Name or Material | Triacetin (Glycerol Triacetate) |