missing translation for 'onlineSavingsMsg'
Learn More

Tri-tert-butylphosphonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7137296
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25841G 1 g
T25845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25841G Supplier TCI America Supplier No. T25841G

Chemical Identifiers

CAS 131274-22-1
Molecular Formula C12H28BF4P
Molecular Weight (g/mol) 290.13
MDL Number MFCD04039975
InChI Key YTJUCJAUJCXFTN-UHFFFAOYSA-O
Synonym tri-tert-butylphosphonium tetrafluoroborate, t-bu 3ph bf4, tri-t-butylphosphonium tetrafluoroborate, fluoroboric acid tri-tert-butylphosphine adduct, tri-tert-butylphosphanium tetrafluoroborate, tbu3p hbf4, ttbp hbf4, t-bu3p hbf4, tbu 3p hbf4, acmc-20a4f9
PubChem CID 2734635
IUPAC Name tetrafluoroboranuide; tri-tert-butylphosphanium
SMILES F[B-](F)(F)F.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C

Specifications

Melting Point 261°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 290.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tri-tert-butylphosphonium Tetrafluoroborate