missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Hydrogen Phosphate Heptahydrate, ACS, EMSURE™, MilliporeSigma™

Catalog No. m1065751000
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065751000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065751000 Supplier Technidata Supplier No. 1.06575.1000
Only null left

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 48°C
pH 8.7 - 9.3
Quantity 1 kg
Solubility Information 154 g/L
Formula Weight 268.03
Packaging Plastic Bottle
Density 1.68 g/cm3 (20 °C); Bulk Density: 800 kg/m3
Assay 98.0 - 102.0%
Chemical Name or Material di-Sodium Hydrogen Phosphate Heptahydrate
RTECSNumber WC4600000
storageNote1 Store at +5°C to +30°C.