missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent

Catalog No. PI49001
Encompass
Change view
Click to view available options
Quantity:
10 x 1 mL ampules
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI49001 50 mL
PI48999 10 x 1 mL ampules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI49001 Supplier Thermo Scientific™ Supplier No. TS49001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent,

Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent, Composed of HMDS, TMCS and high-purity pyridine, it rapidly produces TMS derivatives of polar compounds for GC analysis and biochemical synthesis. This versatile reagent is ideally suited for GC determinations of a range of compounds including sugars, alcohols, phenols, steroids, sterols, organic acids, and some amines.

  • Derivatizes carbohydrates, phenols, steroids, sterols, organic acids, alcohols and some amines
  • Useful for rapid production of TMS derivatives of polar compounds for gas chromatographic determination and biochemical synthesis
  • Versatility, speed and ease of use of Tri-Suil HTP Reagent has made it the most widely used silylation formulation

Specifications

For Use With GC
Standard Concentration 65 - 80, 10 - 20, 5 - 7%
Hazards Corrosive, Flammable
Storage 2°C to 8°C
Quantity 50 mL
Standard Components Pyridine, Hexamethyldisilazane, Chlorotrimethylsilane

For Research Use Only. Not for use in diagnostic procedures.